Learn More
Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Controls
Description
Ensure accurate results when testing with the newest TDM reagents with Thermo Scientific™ QMS™ TDM Controls, offered in three levels.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Lamotrigine Controls |
| Concentration | 2.0, 15.0, 25.0 μg/mL |
| Detectable Analytes | Lamotrigine |
| Sample Type | Plasma, Serum |
| Type | Control |
| Level | 3 |
| Quantity | 3 x 2.5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.