Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Assays
GSA_VA

Catalog No. 23278242 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 x 14 mL, 1 x 12 mL
1 x 19 mL, 1 x 14 mL
1 x 19 mL, 1 x 7 mL
1 x 19 mL, 1 x 8 mL
1 x 22 mL, 1 x 16 mL
1 x 22 mL, 1 x 9 mL
2 x 19 mL
2 x 19 mL, 2 x 7 mL
2 x 19 mL, 2 x 8 mL
2 x 22 mL
2 x 22 mL, 2 x 8 mL
Detectable Analytes:
Amikacin
Gentamicin
Gentamicin (Roche customers, please contact your Roche Sales Representative to purchase this kit)
Lamotrigine
Quinidine
Tobramycin
Topiramate
Vancomycin
Zonisamide
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Detectable Analytes
23-278-242 2 x 19 mL, 2 x 7 mL Amikacin
21-200-419 1 x 22 mL, 1 x 9 mL Gentamicin (Roche customers, please contact your Roche Sales Representative to purchase this kit)
23-279-102 2 x 19 mL Lamotrigine
23-279-101 2 x 19 mL, 2 x 7 mL Quinidine
23-279-103 1 x 22 mL, 1 x 16 mL Topiramate
21-200-440 2 x 22 mL Vancomycin
23-279-100 2 x 22 mL, 2 x 8 mL Zonisamide
23-278-451 1 x 19 mL, 1 x 7 mL Amikacin
23-278-449 1 x 19 mL, 1 x 8 mL Gentamicin
23-278-450 1 x 14 mL, 1 x 12 mL Tobramycin
23-278-453 1 x 19 mL, 1 x 14 mL Topiramate
23-278-454 2 x 19 mL Vancomycin
23-278-455 2 x 19 mL, 2 x 8 mL Zonisamide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 23-278-242 Supplier Thermo Scientific™ Supplier No. 0373910
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Utilize the precision, consistency and optimized performance of Thermo Scientific™ QMS™ TDM Assays, ready-to-use, liquid immunoassays.

Accurately measure therapeutic drug levels in serum and plasma with the Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Assays. These liquid, ready-to-use assays are a convenient alternative to more expensive and time-consuming technologies. QMS (Quantitative Microsphere System) immunoassays have excellent precision and accuracy, optimized performance on a wide range of general chemistry analyzers and lot-to-lot consistency.

Reduces Labor
  • Significantly reduces labor, time, and expense compared to HPLC or LCMS and other technologies
  • Based on competitive inhibition immunoassay principle whereby free-drug in the sample competes for antibody binding sites with drug molecules coated onto uniform microparticles
Ready-to-use
  • Liquid stable and ready-to-use
  • Applicable to general chemistry analyzers
  • Validated parameters assure lab-to-lab reproducibility
  • Superb reagent stability
Optimal Sensitivity
  • Excellent correlation to reference methods
  • Optimal sensitivity, precision and dynamic range
  • Low cross-reactivity with metabolites and commonly co-administered drugs
  • Low interference from endogenous substances

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Amikacin Assay
Detectable Analytes Amikacin
Sample Type Plasma, Serum
Type Reagent
Quantity 2 x 19 mL, 2 x 7 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.