Learn More
Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Calibrators
Description
Accurately measure therapeutic drug levels in serum and plasma with Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Calibrators, which employ advanced microparticle technology that is easy-to-use.
- Significantly reduces labor, time, and expense compared to HPLC or LCMS and other technologies
- Based on competitive inhibition immunoassay principle whereby free-drug in the sample competes for antibody binding sites with drug molecules coated onto uniform microparticles
- Liquid stable and ready-to-use
- Applicable to general chemistry analyzers
- Validated parameters assure lab-to-lab reproducibility
- Superb reagent stability
- Excellent correlation to reference methods
- Low cross-reactivity with metabolites and commonly co-administered drugs
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Detectable Analytes | Gentamicin |
| Sample Type | Plasma, Serum |
| Type | Reagent |
| Quantity | Levels A-F, 1 x 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.