Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ White Polypropylene SturdeeSeal PE Foam Lined Caps

Catalog No. 02991837
Change view
Click to view available options
For Use With (Equipment):
Bottle
Wide mouth bottle
Neck Finish:
120-400
20-410
24-410
28-410
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
89-400
No. per Case:
144 Cs.
1500 Cs.
1650 Cs.
2000 Cs.
210 Cs.
2500 Cs.
2700 Cs.
288 Cs.
3000 Cs.
3750 Cs.
4300 Cs.
5000 Cs.
576 Cs.
608 Cs.
7200 Cs.
760 Cs.
Packaging:
Bagged
Bulk
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Neck Finish No. per Case Packaging
02-991-837 Wide mouth bottle 38-400 576 Cs. Bagged
02-991-013 Wide mouth bottle 120-400 210 Cs. Bulk
02-991-068 Wide mouth bottle 89-400 608 Cs. Bulk
02-991-142 Wide mouth bottle 43-400 3000 Cs. Bulk
02-991-171 Wide mouth bottle 70-400 760 Cs. Bulk
02-991-194 Wide mouth bottle 53-400 2000 Cs. Bulk
02-991-269 Bottle 24-410 4300 Cs. Bulk
02-991-271 Wide mouth bottle 33-400 5000 Cs. Bulk
02-991-270 Bottle 28-410 2700 Cs. Bulk
02-991-273 Wide mouth bottle 63-400 1500 Cs. Bulk
02-991-272 Wide mouth bottle 38-400 3750 Cs. Bulk
02-991-343 Wide mouth bottle 48-400 2500 Cs. Bulk
02-991-440 Bottle 20-410 7200 Cs. Bulk
02-991-443 Wide mouth bottle 45-400 2000 Cs. Bulk
02-991-143 Wide mouth bottle 58-400 1650 Cs. Bulk
02-991-410 Wide mouth bottle 89-400 144 Cs. Bagged
02-991-426 Wide mouth bottle 53-400 288 Cs. Bagged
02-991-541 Wide mouth bottle 33-400 576 Cs. Bagged
02-991-731 Wide mouth bottle 43-400 576 Cs. Bagged
02-991-918 Bottle 24-410 576 Cs. Bagged
02-992-741 Wide mouth bottle 48-400 576 Cs. Bagged
02-992-823 Wide mouth bottle 45-400 576 Cs. Bagged
02-992-847 Wide mouth bottle 58-400 288 Cs. Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Catalog No. 02-991-837 Supplier Qorpak™ Supplier No. CAP06625

Features excellent sealing properties

  • Can be used when good impact strength and pliability are required
  • Resistant to many chemicals including acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents
  • Available in ribbed style
  • Available in bulk or bagged packing

Specifications

Color White
Material Polypropylene
No. per Case 576 Cs.
No. per Pack 12/Pk.
Packaging Bagged
Sterility Non-sterile
Neck Finish 38-400
Disposable Reusable
Product Type Threaded Screw Cap
For Use With (Equipment) Wide mouth bottle
Closure Liner Polyethylene Foam
Closure Type Ribbed
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.