Learn More
Qorpak™ Alpha Caps for F-Style Cans
Description
- Impact resistant and shows wide range of temperature tolerances
- Available with pulp/alum foil or Solvseal liner
- Solvseal liners are ideal for vegetable and mineral oils, weak acids and alkalis, alcohols and organic solvents
- Aluminum foil lined caps are used for food applications and can be used for non-acid, non-alkaline products
Specifications
Specifications
| Color | Silver |
| Material | Metal |
| No. per Case | 2700 |
| Sterility | Non-sterile |
| Liner Material | Solvseal |
| Disposable | Reusable |
| Product Type | Threaded Screw Cap |
| For Use With (Equipment) | Vegetable and mineral oil |
| Autoclavable | Not Autoclavable |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.