Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Borosilicate Glass Crimp Neck Serum Vials without Caps

Catalog No. 02991210
Change view
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
Color:
Amber
Clear
Volume (English):
0.5 dr.
1.4 dr.
2.7 dr.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Volume (English)
02-991-210 2 mL Clear 0.5 dr.
02-991-102 5 mL Amber 1.4 dr.
02-991-196 10 mL Amber 2.7 dr.
02-991-244 10 mL Clear 2.7 dr.
02-991-290 5 mL Clear 1.4 dr.
02-991-214 2 mL Amber 0.5 dr.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 02-991-210 Supplier Qorpak™ Supplier No. 235277

Ideal for pharmaceutical packaging, lyophilization, and sample storage

  • Break resistant and typically used for handling and containment of pharmaceuticals and serums
  • Suitablel for packing and storing blood plasma, media, reagents and vaccines
  • Available in two colors; clear for maximum visibility and amber for UV protection

Specifications

Color Clear
Diameter (Metric) Outer 14.75 mm
Material Borosilicate Glass
Pre-slit No
Sterility Non-sterile
Volume (Metric) 2 mL
For Use With (Application) Ideal for pharmaceutical packaging, lyophilization, and sample storage
Thread Style Crimp
Volume (English) 0.5 dr.
Closure Type Screw Cap
Quantity 4950/Cs.
Marking Spot No
Autoclavable Autoclavable
Height (Metric) 32 mm
Product Type Serum Vial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.