Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Clear Graduated Medium Round Ready to Clean Bottles

Catalog No. 02991072
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
120 mL
240 mL
Packaging:
Carton pack
Conventional Pack
Quantity:
24/Cs.
48/Cs.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Packaging Quantity
02-991-072 30 mL Conventional Pack 48/Cs.
02-992-160 60 mL Conventional Pack 48/Cs.
02-992-172 120 mL Carton pack 24/Cs.
02-992-163 240 mL Conventional Pack 24/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-991-072 Supplier Qorpak™ Supplier No. GLC10523

With slight shoulder for easy pouring and handling

  • Made of glass
  • Wide mouth
  • Graduated in both milliliters and ounces
  • Multipurpose containers for use with biological and pathological specimens and for storing industrial laboratory chemicals and reagents
  • With black polypropylene caps and PTFE discs
  • Available in volumes ranging from 1 to 8 oz. (30 to 240mL)

Specifications

Capacity 1 oz., 30mL
Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Clear
Diameter (English) 1.34 in.
Material Glass
Packaging Conventional Pack
Quantity 48/Cs.
Sterility Non-sterile
Closure Color Black
Closure Included Yes
Graduated Yes
Mouth Wide, Threaded
Shape Round
Height (Metric) 68 mm
Product Type Graduated Medium Round Bottle
Height (English) 2.68 in.
Closure Liner PTFE
Closure Material Polypropylene
Closure Size 33-400 mm
Diameter (Metric) 34 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.