Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Graduated Medium Round Bottles — With Metal Pulp/PE Cap

Catalog No. 02992665
Change view
Click to view available options
Capacity (Metric):
60 mL
120 mL
Closure Size:
38-400 mm
48-400 mm
Packaging:
Bulk Pack
Conventional Pack
Quantity:
24/Cs.
288/Cs.
48/Cs.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Size Packaging Quantity
02-992-665 60 mL 38-400 mm Conventional Pack 48/Cs.
02-991-499 60 mL 38-400 mm Bulk Pack 288/Cs.
02-992-557 120 mL 48-400 mm Conventional Pack 24/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-992-665 Supplier Qorpak™ Supplier No. GLC01481

Mix, measure and store in the same container

  • Designed with slight shoulder for easy pouring and handling
  • Molded graduations in mL and ounces
  • Made of clear glass
  • For use with biological and pathological specimens
  • For storage of industrial laboratory chemicals and reagents

Order Info

Bottles with metal plastisol cap also available (Cat. No. 02-992-180).

Specifications

Capacity 2 oz., 60 mL
Capacity (Metric) 60 mL
Capacity (English) 2 oz.
Color Clear
Diameter (English) 1.65 in.
Material Glass
Packaging Conventional Pack
Quantity 48/Cs.
Closure Color White
Height (Metric) 83 mm
Height (English) 3.27 in.
Closure Liner Pulp/Polyethylene
Closure Size 38-400 mm
Diameter (Metric) 42 mm
Closure Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.