Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Phenolic and Polypropylene Hole Caps

Catalog No. 02991126
Change view
Click to view available options
Neck Finish:
13-425
15-425
18-400
20-400
22-400
24-400
24-410
Material:
Phenolic Resin
Polypropylene
No. per Case:
100 Cs.
10500 Cs.
17000 Cs.
3700 Cs.
4000 Cs.
500 Cs.
5300 Cs.
576 Cs.
5800 Cs.
7500 Cs.
Packaging:
Bagged
Bulk
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Material No. per Case Packaging
02-991-126 15-425 Phenolic Resin 10500 Cs. Bulk
02-991-061 22-400 Phenolic Resin 5800 Cs. Bulk
02-991-127 24-410 Polypropylene 3700 Cs. Bulk
02-991-125 13-425 Phenolic Resin 17000 Cs. Bulk
02-991-188 24-400 Phenolic Resin 4000 Cs. Bulk
02-991-252 20-400 Phenolic Resin 5300 Cs. Bulk
02-991-430 18-400 Phenolic Resin 7500 Cs. Bulk
02-993-084 24-410 Polypropylene 576 Cs. Bagged
02-993-128 24-410 Polypropylene 100 Cs. Bulk
02-993-150 20-400 Phenolic Resin 500 Cs. Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 02-991-126 Supplier Qorpak™ Supplier No. CAP05060
Add to Cart
Add to Cart

Provides good impact strength

Phenolic Hole Caps

  • Pliable
  • Chemically compatible
  • Most temperature tolerant among plastic caps
  • Provide consistent adherence to close dimensional tolerances
  • Ideal for pharmaceutical applications
  • Septa sold separately
Polypropylene Hole Caps
  • Pliable
  • Septa sold separately

Specifications

Color Black
Material Phenolic Resin
No. per Case 10500 Cs.
Packaging Bulk
Sterility Non-sterile
Neck Finish 15-425
Disposable Reusable
Product Type Hole Cap
For Use With (Equipment) Narrow mouth bottle
Autoclavable Not Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.