Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ ValuLine F217 and PTFE Lined Polypropylene Caps

Catalog No. 239081
Change view
Click to view available options
Neck Finish:
20-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
89-400
No. per Case:
10225 Cs.
1100 Cs.
1300 Cs.
1700 Cs.
2000 Cs.
2200 Cs.
2900 Cs.
4000 Cs.
456 Cs.
5000 Cs.
6500 Cs.
760 Cs.
900 Cs.
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case
23-908-1 33-400 4000 Cs.
23-908-0 28-400 5000 Cs.
23-908-4 45-400 2000 Cs.
23-908-5 48-400 1700 Cs.
23-908-6 53-400 1300 Cs.
23-908-2 38-400 2900 Cs.
23-908-7 58-400 1100 Cs.
23-908-8 63-400 900 Cs.
23-908-3 43-400 2200 Cs.
23-908-9 70-400 760 Cs.
23-909-0 89-400 456 Cs.
02-991-312 24-400 6500 Cs.
02-991-542 20-400 10225 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 23-908-1 Supplier Qorpak™ Supplier No. 239081
Add to Cart
Add to Cart

Help to keep bottle or jar contents free of odor and taste interference

  • Made of white polypropylene with PTFE facing
  • F217 liner backing consisting of low density foam core tightly sandwiched between two layers of LDPE
  • Resilient with strong resistance to chemicals and moisture

Specifications

Color White
Material Polypropylene
No. per Case 4000 Cs.
Packaging Bulk
Sterility Non-sterile
Liner Material F217, PTFE
Neck Finish 33-400
Disposable Reusable
Product Type Lined Cap (Ribbed)
For Use With (Equipment) Wide mouth bottle
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.