Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ White Metal Pulp/Aluminum Foil Lined Caps

Catalog No. 02993382
Change view
Click to view available options
Neck Finish:
24-400
38-400
43-400
48-400
58-400
63-400
70-400
89-400
No. per Case:
144 Cs.
288 Cs.
3000 Cs.
576 Cs.
800 Cs.
Packaging:
Bagged
Bulk
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case Packaging
02-993-382 58-400 288 Cs. Bagged
02-991-111 89-400 800 Cs. Bulk
02-991-253 38-400 3000 Cs. Bulk
02-993-368 43-400 576 Cs. Bagged
02-993-399 89-400 144 Cs. Bagged
02-993-418 48-400 576 Cs. Bagged
02-993-366 24-400 576 Cs. Bagged
02-993-370 63-400 288 Cs. Bagged
02-993-391 38-400 576 Cs. Bagged
02-993-417 70-400 144 Cs. Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 02-993-382 Supplier Qorpak™ Supplier No. CAP06653
Add to Cart
Add to Cart

Free of odor and taste problems

  • Made of steel coated with anti-corrosive tinplate
  • Impact resistant
  • Tolerate wide ranges of temperature
  • Feature standard 400 GPI thread finish
  • Used for non-acid, non-alkaline products, organic solvents, chrome cleaners, food bottles and jars, brake fluids and mineral oils

Specifications

Color White
Material Metal
No. per Case 288 Cs.
No. per Pack 12/Pk.
Packaging Bagged
Sterility Non-sterile
Liner Material Pulp/Aluminum Foil
Neck Finish 58-400
Disposable Reusable
Product Type Threaded Screw Cap
For Use With (Equipment) Wide mouth bottle
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.