Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ White Polypropylene Flip Top Caps

Catalog No. 02992709
Change view
Click to view available options
For Use With (Equipment):
Bottle
Wide mouth bottle
Neck Finish:
24-410
28-400
No. per Case:
144 Cs.
1800 Cs.
1880 Cs.
Packaging:
Bagged
Bulk
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Neck Finish No. per Case Packaging
02-992-709 Bottle 24-410 144 Cs. Bagged
02-991-124 Wide mouth bottle 28-400 1800 Cs. Bulk
02-991-177 Bottle 24-410 1880 Cs. Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 02-992-709 Supplier Qorpak™ Supplier No. CAP06047

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Ideal on workbench for dispensing small amounts of liquid

  • Made of durable polypropylene
  • Unlined
  • Cap orifice: 0.11 in. (3 mm)

Specifications

Color White
Material Polypropylene
No. per Case 144 Cs.
Packaging Bagged
Sterility Non-sterile
Neck Finish 24-410
Disposable Reusable
Product Type Threaded Screw Cap
For Use With (Equipment) Bottle
Autoclavable Not Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.