Learn More
QLAB CORPORATION QPANEL TEST SUBSTRATE A-2X3.5
SDP
Supplier:  QLAB CORPORATION A23.5
Encompass
Q-PANEL® aluminum test substrates from Q-Lab minimize metal variability as a source of bias in critical tests. Made from high-quality aluminum, they are clean, consistent, convenient, and economical. Sold per box of 500 panels.
Catalog No. NC2187947
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.