Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QPCT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576997 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576997 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576997 Supplier Invitrogen™ Supplier No. PA576997
  • $684.00
    $525.65 / Each of 1

    Save $158.35

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

GPIP137 regulates the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation/migration in multiple cell types. It binds directly and selectively to MYC and CCND2 RNAs. In neuronal cells, GPIP137 binds to several mRNAs associated with RNA granules including BDNF, CAMK2A, CREB1, MAP2, NTRK2, mRNAs, GRIN1, and KPNB1 mRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen QPCT
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Qpct
Gene Accession No. Q16769, Q9CYK2
Gene Alias 5730422A13Rik; EC; GCT; Glutaminyl cyclase; glutaminyl-peptide cyclotransferase; glutaminyl-peptide cyclotransferase (glutaminyl cyclase); glutaminyl-tRNA cyclotransferase; Glutamyl cyclase; QC; Qpct; RGD1562284; similar to Glutaminyl-peptide cyclotransferase precursor (QC); sQC
Gene Symbols Qpct
Host Species Rabbit
Immunogen Recombinant full length Human QPCT.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25797, 313837, 70536
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.