Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QRP™ Qualatex™ 5C Powder-Free Latex Finger Cots

Catalog No. 19048700B Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Cleanroom Class:
Class 100 (ISO 5)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cleanroom Class
19-048-700B Class 100 (ISO 5)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-048-700B Supplier QRP™ Supplier No. 5CM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Eliminates contamination risks from silicone oils, skin salts, particulates, and plasticizers. Packaged for ISO 5 (Class 100) cleanroom applications in a unique standup pouch for easy product identification.

  • Eliminates contamination risks caused by silicone oils, skin salts, particulates, and plasticizers
  • Packaged for ISO 5 (Class 100) cleanroom applications
  • Compliant with ASTM and IEST-RP-CC 005.3 standards
  • Unique, standup pouch allows for easy product identification

Specifications

Certifications/Compliance ASTM/IEST-RP-CC 005.3
Chlorinated Yes
Cleanroom Class Class 100 (ISO 5)
Accelerator Free Yes
Applications Laboratory
Color Natural
Disposable Single-use
Hand Ambidextrous
Latex Free No
Material Latex
Powdered Powder Free
Product Type Finger Cot
Size Medium
Sterility Sterile
Thickness (English) 3.5 mil
Thickness (Metric) 0.09 mm
For Use With (Application) Discreet Components, Food Processing, Hard Drive Assembly, and Optics
Glove Cuff Style Open
Packaging Type Stand-up Pouch
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.