Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qsonica Q700/Q500 Accessories: 8 Tube Holder Rack
SDP

An excellent solution for securely holding tubes in place when working with Q500/Q700 Sonicators.

$236.00 - $245.00

Specifications

Product Type Tube Holder Rack
For Use With (Equipment) Cup Horn (Part # 431C2)
View More Specs

Products 2
Catalog Number Mfr. No. Holds Color Price Quantity & Availability  
Catalog Number Mfr. No. Holds Color Price Quantity & Availability  
15-338-323
SDP
View Documents
Qsonica
440
8 x 1.5 mL Polystyrene Evergreen Tubes White
Each for $245.00
Only null left
Add to Cart
 
15-338-324
SDP
View Documents
Qsonica
451
8 x 1.5 mL Polypropylene Eppendorf Tubes Green
Each for $236.00
Only null left
Add to Cart
 
Description

Description

  • Holds 8 tubes of 1.5mL
  • Available for either Evergreen or Eppendorf tubes
Specifications

Specifications

Tube Holder Rack
Cup Horn (Part # 431C2)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.