Learn More
Medchemexpress LLC Qtz | 2883232-39-9 | 96.0% | 428.48 g·mol⁻¹ | C28H20N4O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY4717610MG
Encompass_Preferred
QTZ is a coelenterazine-analog bioluminescence agent for in vivo imaging. It produces red-shifted emission with low background, enabling sensitive luciferase-based live-animal imaging. Supplied as a powder or as a DMSO solution, it is intended for laboratory use under appropriate handling and storage conditions.
  • Red-shifted bioluminescent emission with low background
  • Suitable for luciferase-based in vivo imaging
  • Available as powder or ready-to-use DMSO solution
  • Multiple pack sizes to support small- and large-scale experiments
  • Stable under frozen storage; follow manufacturer storage recommendations
Catalog No. 50-002-97950