Learn More
Binder Inc Qualification documents
SDP
Supplier:  Binder Inc 70570001
Encompass_Preferred
Qualification documents - Digital in PDF format, IQ/OQ documents – supporting documents for validation performed by customers, consisting of: IQ/OQ checklists incl. calibration guide and comprehensive unit documentation; parameters: temperature, CO2, O2, pressure, depending on unit
Catalog No. 70-570-001
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.