Learn More
MilliporeSigma™ Qualification Workbook
Provides instructions on performing system qual and completing qual forms
Supplier:  MilliporeSigma™ VFREXP350
 View more versions of this product
Catalog No. VFREXP350
Only null left
Add to Cart
Includes:
IQ and OQ protocols, recommended PQ protocols and annual system maintenance procedures
Description
Description
  • For use with Milli-RO, Milli-RX, Milli-Q, RiOs, SDS, Elix, Direct-Q systems
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SDS 350 System
Qualification Workbook
Provides instructions on performing system qual and completing qual forms
IQ and OQ protocols, recommended PQ protocols and annual system maintenance procedures
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.