Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Control Standard Mixture, Restek™
SDP

Catalog No. 06802066 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-802-066 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-802-066 Supplier Restek Corporation Supplier No. 31631
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Reference mixtures for ISO/DIS 9377 (German H-53), a gas chromatography flame ionization detection (GC-FID) method.
  • For GC analysis of total petroleum hydrocarbons (TPH) in water.
  • Environmentally safer
  • Calibration standard available as diesel #2/motor oil and diesel #2/mineral oil.
Components:
  • Diesel #2 (additive-free) (68334-30-5)
  • Mineral oil (additive-free [i.e., USP grade]

Specifications

Packaging Ampule
CAS Max % 99.90
Odor Strong
Specific Gravity 0.7845 g/cm3 at 25°C
Molecular Formula C3H6O
Concentration 500 μg/mL each in acetone, Total hydrocarbon concentration is 1000 μg/mL.
MDL Number MFCD00008765
Solubility Completely soluble
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
Quantity 1 mL
Formula Weight 58.08
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.