Learn More
MBL International Corporation QuickSwitch„¢ Quant HLA-DRB1*01:01 Tetramer Kit-PE
SDP
Supplier:  MBL International Corporation TB7500K1
Encompass
Allele:DRB1*01:01; Application:FCM; Conjugate:PE; Peptide Sequence:Custom; Species Reactivity:Human; Size:10 test; Target:Custom; QuickSwitch Class II Tetramer Peptide Exchange and Quantitation Kit utilizes a patented technique for exchanging up to ten peptides on an MHC Class II Tetramer. Reduce time and cost with this kit to create your own custom tetramers or to test which peptide sequence has the best peptide-MHC binding affinity.  This kit can be used for screening multiple peptides or to create a small amount of your own custom Tetramer in house.  The resulting custom tetramers can be used for multiple applications including screening immunogenic peptides, neoantigen discovery, and many other assays downstream from in-silico  selection.FAQs for Peptide Exchange using QuickSwitch Kit
Catalog No. NC2456189
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.