Learn More
Bioassay Systems QuantiChrom™ Copper Assay Kit. For quantitative determination of copper(II) ion and evaluation of drug effects on copper metabolism.
SDP
Catalog No. 50489214 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of copper(II) ion and evaluation of drug effects on Cu metabolism. Key Features: Sensitive and accurate. Linear detection range 7 µg/dL (1.0 µM) to 300 µg/dL (47 µM) copper in 96-well plate assay. Simple and high-throughput. The simple procedure can be readily automated as a high-throughput assay in 96-well plates for thousands of samples per day. Improved reagent stability and versatility. The optimized formulation has greatly enhanced reagent and signal stability. Cuvet or 96-well plate assay. Method: OD359nm. Samples: Biological, environment, food and beverage. Species: All. Procedure: Assay takes 10 min. Kit size: 250 tests. Detection limit: 7 µg/dL (1.0 µM).
Catalog No. 50-489-214 Supplier Bioassay Systems Supplier No. DICU250
For quantitative determination of copper(II) ion and evaluation of drug effects on Cu metabolism. Key Features: Sensitive and accurate. Linear detection range 7 µg/dL (1.0 µM) to 300 µg/dL (47 µM) copper in 96-well plate assay. Simple and high-throughput. The simple procedure can be readily automated as a high-throughput assay in 96-well plates for thousands of samples per day. Improved reagent stability and versatility. The optimized formulation has greatly enhanced reagent and signal stability. Cuvet or 96-well plate assay. Method: OD359nm. Samples: Biological, environment, food and beverage. Species: All. Procedure: Assay takes 10 min. Kit size: 250 tests. Detection limit: 7 µg/dL (1.0 µM).