Learn More
Bioassay Systems QuantiChrom trade; Ethanol Assay Kit. For quantitative determination of ethanol and alcohol metabolism.
SDP
Catalog No. 50489215 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of ethanol and alcohol metabolism. Key Features: Sensitive and accurate. Detection range 0.04 - 4% alcohol in 96-well plate assay. Convenient and high-throughput. The procedure involves adding a single working reagent, incubation for 8 min, adding a Stop Reagent, and reading the optical density. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Versatility. Assays can be executed in 96-well plate or cuvet. Method: OD580nm (Chemical). Samples: Alcoholic beverages etc. Species: All. Procedure: Assay takes 10 min. Kit size: 500 tests. Detection limit: 0.04%.
Catalog No. 50-489-215 Supplier Bioassay Systems Supplier No. DIET500
For quantitative determination of ethanol and alcohol metabolism. Key Features: Sensitive and accurate. Detection range 0.04 - 4% alcohol in 96-well plate assay. Convenient and high-throughput. The procedure involves adding a single working reagent, incubation for 8 min, adding a Stop Reagent, and reading the optical density. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Versatility. Assays can be executed in 96-well plate or cuvet. Method: OD580nm (Chemical). Samples: Alcoholic beverages etc. Species: All. Procedure: Assay takes 10 min. Kit size: 500 tests. Detection limit: 0.04%.