Learn More
Bioassay Systems QuantiChrom™ Glycosaminoglycans Assay Kit, For quantitative determination of glycosaminoglycans in food and tissue homogenate.
SDP
Catalog No. 5000144363 Shop All Bioassay Systems Products
Encompass_Preferred
GLYCOSAMINOGLYCANS (GAGs) are long, linear polysaccharides and highly negatively charged molecules that constitute a major component of the extracellular matrix in mammalian tissues. GAGs play key roles in cell growth, cell adhesion, wound repair, and anticoagulation. They are also important for brain development and homeostasis. GAGs are associated with osteoarthritis, inflammation, diabetes, spinal cord injury, and cancer.BioAssay Systems’ GAGs assay uses a turbidimetric reaction in which GAGs form an insoluble complex with protein in the reagent. The increase in turbidity at 600nm is directly proportional to the glycosaminoglycans in the sample (reported in hyaluronic acid equivalents).
Catalog No. 50-001-44363 Supplier Bioassay Systems Supplier No. DGAG100
Add to Cart
Add to Cart
GLYCOSAMINOGLYCANS (GAGs) are long, linear polysaccharides and highly negatively charged molecules that constitute a major component of the extracellular matrix in mammalian tissues. GAGs play key roles in cell growth, cell adhesion, wound repair, and anticoagulation. They are also important for brain development and homeostasis. GAGs are associated with osteoarthritis, inflammation, diabetes, spinal cord injury, and cancer.BioAssay Systems’ GAGs assay uses a turbidimetric reaction in which GAGs form an insoluble complex with protein in the reagent. The increase in turbidity at 600nm is directly proportional to the glycosaminoglycans in the sample (reported in hyaluronic acid equivalents).