Learn More
Bioassay Systems QuantiChrom™ Heme Assay Kit. For quantitative determination of heme and evaluation of drug effects on heme metabolism.
SDP
Catalog No. 50489220 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of heme and evaluation of drug effects on heme metabolism. Key Features: Sensitive and accurate. Linear detection range 0.6 - 125 µM heme in 96-well plate assay. Simple and high-throughput. The "mix-and-read" procedure involves addition of a single working reagent and reading the optical density. Can be readily automated as a high-throughput assay in 96-well plates for thousands of samples per day. Safety. Reagents are non-toxic. Versatility. Assays can be executed in 96-well plate or cuvet. Method: OD400nm. Samples: Blood, serum, plasma, urine etc. Species: All. Procedure: Assay takes 5 min. Kit size: 250 tests. Detection limit: 0.6 µM.
Catalog No. 50-489-220 Supplier Bioassay Systems Supplier No. DIHM250
For quantitative determination of heme and evaluation of drug effects on heme metabolism. Key Features: Sensitive and accurate. Linear detection range 0.6 - 125 µM heme in 96-well plate assay. Simple and high-throughput. The "mix-and-read" procedure involves addition of a single working reagent and reading the optical density. Can be readily automated as a high-throughput assay in 96-well plates for thousands of samples per day. Safety. Reagents are non-toxic. Versatility. Assays can be executed in 96-well plate or cuvet. Method: OD400nm. Samples: Blood, serum, plasma, urine etc. Species: All. Procedure: Assay takes 5 min. Kit size: 250 tests. Detection limit: 0.6 µM.