Learn More
Bioassay Systems QuantiChrom™ Lactate Dehydrogenase Kit. For quantitative determination of lactate dehydrogenase LDH activity and screen/evaluation of LDH modulators.
SDP
Catalog No. NC0920522 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of lactate dehydrogenase LDH activity and screen/evaluation of LDH modulators. Key Features: High sensitivity and wide linear range. Use 3 µL serum or plasma sample. The detection limit is 2 U/L, linear up to 200 U/L. Homogeneous and simple procedure. Simple "mix-and-measure" procedure allows reliable quantitation of LDH activity within 30 minutes. Robust and amenable to HTS. All reagents are compatible with high-throughput liquid handling instruments. Method: OD565nm. Samples: Serum, plasma etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 2 U/L.
Catalog No. NC0920522 Supplier Bioassay Systems Supplier No. D2DH100
For quantitative determination of lactate dehydrogenase LDH activity and screen/evaluation of LDH modulators. Key Features: High sensitivity and wide linear range. Use 3 µL serum or plasma sample. The detection limit is 2 U/L, linear up to 200 U/L. Homogeneous and simple procedure. Simple "mix-and-measure" procedure allows reliable quantitation of LDH activity within 30 minutes. Robust and amenable to HTS. All reagents are compatible with high-throughput liquid handling instruments. Method: OD565nm. Samples: Serum, plasma etc. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 2 U/L.