Learn More
Bioassay Systems QuantiChrom™ TBARS Assay Kit. For quantitative determination of lipid peroxides (thiobarbituric acid reactive substances, TBARS) and evaluation of drug effects on lipid peroxidation.
SDP
Catalog No. 50489234 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of lipid peroxides (thiobarbituric acid reactive substances, TBARS) and evaluation of drug effects on lipid peroxidation. Key Features: Sensitive and accurate. Linear detection range: colorimetric assay 1 - 30 µM, fluorometric assay 0.1 - 1.5 µM MDA. Method: OD535nm, or FL530/550nm. Samples: Serum, plasma, urine, saliva etc. Species: All. Procedure: Assay takes 80 min. Kit size: 100 tests. Detection limit: 0.1 µM.
Catalog No. 50-489-234 Supplier Bioassay Systems Supplier No. DTBA100
For quantitative determination of lipid peroxides (thiobarbituric acid reactive substances, TBARS) and evaluation of drug effects on lipid peroxidation. Key Features: Sensitive and accurate. Linear detection range: colorimetric assay 1 - 30 µM, fluorometric assay 0.1 - 1.5 µM MDA. Method: OD535nm, or FL530/550nm. Samples: Serum, plasma, urine, saliva etc. Species: All. Procedure: Assay takes 80 min. Kit size: 100 tests. Detection limit: 0.1 µM.