Learn More
Bioassay Systems QuantiChrom™ Total Carbohydrate Assay Kit, For quantitative determination of total carbohydrates in food, beverage, and biological samples (e.g. serum, plasma, etc).
SDP
Catalog No. 5000144377 Shop All Bioassay Systems Products
Encompass_Preferred
CARBOHYDRATES are made up of carbon, hydrogen, and oxygen. A carbohydrate may exist in its simplest form, a monosaccharide, or as larger polymers called polysaccharides (e.g. starch, glycogen, and cellulose). Carbohydrates are essential constituents of living things, and in the form of glucose, serve as the main source of energy for metabolism. They are also commonly found in many fruits and vegetables as simple or complex sugars. BioAssay Systems’ QuantiChrom™ Total Carbohydrate Assay Kit first hydrolyzes polysaccharides to monosaccharides, which are then converted to furfural compounds that react with the Detection Reagent. The color intensity of the chromagen, measured at 490 nm, is directly proportional to the total carbohydrate concentration (glucose equivalents) in the sample.
Catalog No. 50-001-44377 Supplier Bioassay Systems Supplier No. TCRB100
CARBOHYDRATES are made up of carbon, hydrogen, and oxygen. A carbohydrate may exist in its simplest form, a monosaccharide, or as larger polymers called polysaccharides (e.g. starch, glycogen, and cellulose). Carbohydrates are essential constituents of living things, and in the form of glucose, serve as the main source of energy for metabolism. They are also commonly found in many fruits and vegetables as simple or complex sugars. BioAssay Systems’ QuantiChrom™ Total Carbohydrate Assay Kit first hydrolyzes polysaccharides to monosaccharides, which are then converted to furfural compounds that react with the Detection Reagent. The color intensity of the chromagen, measured at 490 nm, is directly proportional to the total carbohydrate concentration (glucose equivalents) in the sample.