Learn More
Bioassay Systems QuantiFluo™ Ammonia Assay Kit. For quantitative determination of ammonia/ammonium concentration by fluorimetry at 360nm/450nm (compatible on handheld fluorimeter catalog# FL360450).
SDP
Catalog No. 50489229 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of ammonia/ammonium and evaluation of drug effects on their metabolism. Key Features: Fast and sensitive. Linear detection range of 0.012 - 1 mM ammonia. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: FL360/450nm. Samples: Biological (eg urine) and environ-mental samples. Species: All. Procedure: Assay takes 15 min. Kit size: 200 tests. Detection limit: 12 µM.
Catalog No. 50-489-229 Supplier Bioassay Systems Supplier No. DNH3200
For quantitative determination of ammonia/ammonium and evaluation of drug effects on their metabolism. Key Features: Fast and sensitive. Linear detection range of 0.012 - 1 mM ammonia. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: FL360/450nm. Samples: Biological (eg urine) and environ-mental samples. Species: All. Procedure: Assay takes 15 min. Kit size: 200 tests. Detection limit: 12 µM.