Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega QuantiFluor™ ONE dsDNA Dye

Catalog No. PRE4891 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRE4891 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRE4891 Supplier Promega Supplier No. E4891
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The QuantiFluor ONE dsDNA Dye is a fluorescent double-stranded DNA-binding dye (504nmEx/531nmEm) prepared for an add-and-read format making sample quantitation easy. This dye enables sensitive quantitation of small amounts of double-stranded DNA.

  • Sensitive, Fluorescent DNA Quantitation
  • Specific quantitation of dsDNA
  • Increased sensitivity for low-concentration samples
  • Simple to use and automation-friendly

Specifications

Content And Storage -30°C to +10°C
Quantity 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.