Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega QuantiFluor™ ONE dsDNA System

Catalog No. PRE4871 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
No. of Reactions:
100
500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
PRE4871 100
PRE4870 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRE4871 Supplier Promega Supplier No. E4871
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Includes

  • QuantiFluor ONE dsDNA Dye
  • QuantiFluor ONE Lambda DNA
  • 1X TE Buffer, pH 7.5

The QuantiFluor(R) ONE dsDNA System enables sensitive quantitation of small amounts of double-stranded DNA (dsDNA). The system contains a fluorescent dsDNA-binding dye (504nm[Ex]/531nm[Em]) developed for use in an add-and-read format for dye and standard.

  • Fluorescent Quantitation of dsDNA with Add-and-Read Convenience
  • Specific quantitation of dsDNA
  • Increased sensitivity over absorbance at 260nm for low-concentration samples
  • Simple to use and automation-friendly

Specifications

Content And Storage Varies Among Components
No. of Reactions 100
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.