Learn More
Mitegen Holey Carbon Grid By Quantifoil, R 1.2/1.3 Hole Pattern, 300 Gold Mesh, 12nm Thickness, For Cryo-EM, 100/Pack
Supplier: Mitegen MCEMQ3100AR1.3
The Quantifoil Holey Carbon Grid provides an ideal, ultra-flat support for biological samples in cryo-electron microscopy (cryo-EM) techniques. This grid features a precise R 1.2/1.3 hole pattern, with 1.2 µm hole diameter and 1.3 µm interspace, applied on a robust 300 gold electron microscopy mesh. The uniform circular holes and a carbon film thickness of about 12 nm ensure optimal particle dispersion and consistent ice thickness, crucial for achieving high-resolution data. Crafted from gold, this mesh offers excellent conductivity and inherent stability. Designed for reliability and precise sample integration, these grids streamline the cryo-EM workflow for vitrification and subsequent imaging. They are indispensable for obtaining high-quality data and clear visualization, contributing to enhanced resolution and reproducibility in structural biology research. This product is supplied in a convenient 100-pack, providing ample supply for demanding experimental needs.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.