Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QuantiGene™ Homogenizing Solution

Catalog No. QG0516
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 mL
50 mL
200 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QG0516 50 mL
QS0518 20 mL
QG0517 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. QG0516 Supplier Invitrogen™ Supplier No. QG0516
Add to Cart
Add to Cart

QuantiGene Homogenizing Solution is used to prepare tissue homogenates from fresh, frozen, or formalin-fixed paraffin-embedded (FFPE) tissues.

QuantiGene Homogenizing Solution is used to prepare tissue homogenates from fresh, frozen, or formalin-fixed paraffin-embedded (FFPE) tissues. Homogenization of tissues using QuantiGene Homogenizing Solution releases RNA for detection and quantitation with QuantiGene assays. The homogenizing solution is one of two components in the QuantiGene Sample Processing Kit for tissues, and is used to supplement this kit when extra volume is needed.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
For Use With (Application) Multi-Gene Expression
Product Line QuantiGene
Product Type Cell Lysis Buffer
Quantity 50 mL
Shipping Condition Blue Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.