Learn More
Biolegend Biotin anti-human PLAUR uPAR
Supplier: Biolegend 631453
Biotin anti-human PLAUR (uPAR) [A20095T]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: Direct ELISA; Size: 50 μg
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.