Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QuantiGene™ Plex Plate Seals

Catalog No. QG0524
Encompass_Preferred
Change view
Click to view available options
Quantity:
45 seals
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QG0524 45 seals
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QG0524 Supplier Invitrogen™ Supplier No. QG0524
Add to Cart
Add to Cart

QuantiGene Plex Plate Seals are clear microtiter plate seals for use with QuantiGene Plex magnetic separation plates (96- and 384-well).

QuantiGene Plex Plate Seals are clear microtiter plate seals for use with QuantiGene Plex magnetic separation plates (96- and 384-well). These seals are intended to be used only during the branched DNA reagent hybridizations and SAPE incubation on the second day of the QuantiGene Plex Assay.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Quantity 45 seals
Content And Storage 45 clear adhesive plate seals
For Use With (Equipment) QuantiGene Plex Magnetic Separation Plates
Shipping Condition Room Temperature
Product Type Adhesive Seal
Product Line QuantiGene
Plate Compatibility 96/384-well Magnetic Separation Plates

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.