Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ QuantiGene™ Plex Pressure Seals
Description
QuantiGene Plex Pressure Seals are clear, pressure-activated microplate seals for use with 96- and 384-well hybridization plates during the overnight hybridization of the QuantiGene Plex Assay.
Specifications
Specifications
| Color | Clear |
| No. of Wells | 96 |
| Quantity | 12 Seals |
| Content And Storage | 12 pressure-activated seals |
| Shipping Condition | Room Temperature |
| Product Type | Pressure-activated Microplate Seals |
| Product Line | QuantiGene |
| Plate Compatibility | 96/364-well Hybridization Plates |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?