Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuantiGene™ Plex Wash Buffer Components

Catalog No. QP1502
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Plate
10 Plates
3 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QP1502 3 Plates
QP1501 1 Plate
QP1503 10 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. QP1502 Supplier Invitrogen Supplier No. QP1502
Add to Cart
Add to Cart

QuantiGene Plex Wash Buffer Components are aqueous buffered solutions that are mixed with nuclease-free water to make QuantiGene Plex wash buffer.

QuantiGene Plex Wash Buffer Components are aqueous buffered solutions that are mixed with nuclease-free water to make QuantiGene Plex wash buffer. The wash buffer components may be purchased in 1, 3, or 10-plate quantities to supplement QuantiGene Plex Assay kits when additional wash buffer volume is required for automation.
TRUSTED_SUSTAINABILITY

Specifications

Format 96-well
For Use With (Application) QuantiGene Plex Assay
Content And Storage • 2.4 mL Wash Buffer Component 1
• 39 mL Wash Buffer Component 2

Store at room temperature.

Key Functions Multi-Gene Expression
Product Line QuantiGene Plex
Product Type Wash Buffer
Quantity 3 Plates
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.