Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QuantiGene™ Singleplex Foil Plate Seals

Catalog No. QS0506
Encompass_Preferred
Change view
Click to view available options
Quantity:
12 Seals
260 Seals
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QS0506 12 Seals
QS0507 260 Seals
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. QS0506 Supplier Invitrogen™ Supplier No. QS0506
Add to Cart
Add to Cart

QuantiGene Singleplex Foil Plate Seals are intended for use with all hybridization steps of the QuantiGene Singleplex Assay.

QuantiGene Singleplex Foil Plate Seals are intended for use with all hybridization steps of the QuantiGene Singleplex Assay. The foil plate seals are compatible with 96- and 384-well plates.

TRUSTED_SUSTAINABILITY

Specifications

No. of Wells 96
Quantity 12 Seals
Content And Storage 12 foil-backed adhesive plate seals
Shipping Condition Room Temperature
Product Type Foil-Backed Adhesive Seal
Product Line QuantiGene
Plate Compatibility 96/384-well Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.