Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ QuantiGene™ Singleplex Foil Plate Seals
Description
QuantiGene Singleplex Foil Plate Seals are intended for use with all hybridization steps of the QuantiGene Singleplex Assay. The foil plate seals are compatible with 96- and 384-well plates.
Specifications
Specifications
| No. of Wells | 96 |
| Quantity | 260 Seals |
| Content And Storage | 260 foil-backed adhesive plate seals |
| Shipping Condition | Room Temperature |
| Product Type | Foil-Backed Adhesive Seal |
| Product Line | QuantiGene |
| Plate Compatibility | 96/384-well Plate |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?