Learn More
Description
- Programmed Death Ligand 1
- PD-L1, also known as B7-H1 and CD274, is an approximately 65 kDa transmembrane glycoprotein in the B7 family of immune regulatory molecules.
- PD-L1 is expressed on inflammatory-activated immune cells including macrophages, T cells, and B cells, keratinocytes, enothelial and intestinal epithelial cells, as well as a variety of carcinomas and melanoma.
- PD-L1 binds to T cell B7-1/CD80 and PD-1.
- It suppresses T cell activation and proliferation and induces the apoptosis of activated T cells.
- It plays a role in the development of immune tolerance by promoting T cell anergy and enhancing regulatory T cell development.
- PD-L1 favors the development of anti-inflammatory IL-10 and IL-22 producing dendritic cells and inhibits the development of Th17 cells.
- In cancer, PD-L1 provides resistance to T cell mediated lysis, enhances EMT, and enhances the tumorigenic function of Th22 cells.
Specifications
Specifications
| Product Type | ELISA |
| For Use With (Application) | Control |
| For Use With (Equipment) | Catalog number DB7H10 |
| Gene ID (Entrez) | 29126 |
| Host Species | Human, Cynomolgus Monkey |
| Quantity | 1 Ea. |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 |
| Target | PD-L1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
