Learn More
Description
TIM-1 (T cell immunoglobulin and mucin domain 1), also known as KIM-1 and HAVcr1, is expressed on many immune cell types, epithelial cells. It binds to phosphatidylserine (PS), LMIR5/CD300b, TIM-1 (homophilic), TIM-4, IgA, glycoproteins of number of enveloped viruses. Its interaction with PS enables TIM-1 to mediate phagocytosis, clearance of apoptotic cells. TIM-1 signaling costimulates T cell activation, enhances Th2 cytokine production. TIM-1 serves as cellular entry receptor for hepatitis A virus, Ebolavirus and Marburgvirus. Polymorphisms are associated with susceptibility to atopy, autoimmunity, severe hepatitis A virus infection in humans. A soluble form of TIM-1 is elevated in urine during nephropathy.
- Storage: Store unopened product at 2 to 8°C. Do not use past expiration date.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Type | Control Set for ELISA Kit |
| Gene ID (Entrez) | 26762 (Human); 171283 (Mouse); 286934 (Rat); 102141332 (Cynomolgus Monkey) |
| Intended Use | For quantitative measurement of T cell immunoglobulin protein levels |
| Product Line | R&D Systems |
| Product Name | Immunoassay Control Set 920 |
| Source | Human |
| Target Gene | T Cell Immunoglobulin Mucin-1 |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
