Learn More
Description
For use as quantitative controls for the determination of cytokine concentrations in biological fluids. Concentrations have been assigned using R&D Systems™’ Quantikine™ ELISA kits. Controls are prepared in a buffered protein base with preservatives. They contain recombinant human cytokines at low, medium and high concentrations. Controls are supplied lyophilized.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2-8°C. Do not use past expiration date. |
| Gene ID (Entrez) | 3791 |
| Product Line | R&D Systems |
| Quantity | 1 Each |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
