Learn More
R&D Systems™ Quantikine Immunoassay Control Set 1003 for Mouse HS IL-1b
Shop All R&D Systems Products

Specifications
Specifications
| Gene ID (Entrez) | 3553 |
| Host Species | Mouse |
| Quantity | 1 Ea. |
| Storage Requirements | Store the unopened product at 2 to 8 °C. Do not use past expiration date |
| Synonym | catabolin, IL-1, IL1 beta, IL-1 beta, IL1B, IL-1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta |
| Target | IL-1 beta/IL-1F2 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.