Learn More
Description
Interleukin 18 (IL-18), also known as interferon-gamma-inducing factor (IGIF) and IL-1γ, is cytokine which shares biologic activities with IL-12 and structural similarities with IL-1 family of proteins. IL-18 was originally cloned from liver cells, has since been shown to be expressed by monocyte/macrophages, osteoblasts and keratinocytes. Caspase-1 (IL-1 beta-converting enzyme) has been implicated in physiological processing of pro-IL-18 to IL-18. Similarly to IL-12, human IL-18 has been shown to enhance NK cell activity in PBMC cultures. Human IL-18 has also been found to induce production IFN-γ and GM-CSF while inhibiting production of IL-10 by PBMC. On enriched human T cells, human IL-18 can enhance Th1 cytokine production and stimulate cell proliferation via IL-2-dependent pathway. In mouse system, IL-18 has been shown to be costimulatory factor for activation of Th1, but not Th2, cells. IL-18 was found to selectively enhance FasL-mediated cytotoxicity of Th1, but not Th0 or Th2, cells. IL-18 has also been shown to induce activated B cells to produce IFN-γ that inhibits IgE production.
- Storage: Store unopened product at 2 to 8°C. Do not use past expiration date.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Type | Control Set for ELISA Kit |
| Gene ID (Entrez) | 3606 (Human); 16173 (Mouse); 29197 (Rat); 397057 (Porcine); 574151 (Primate) |
| Intended Use | For quantitative measurement of Interleukin 18 protein levels |
| Product Line | R&D Systems |
| Product Name | Immunoassay Control Set 919 |
| Source | Human |
| Target Gene | Interleukin 18 |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
