Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 822 Human Total PAI-1
SDP

Catalog No. QC209 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QC209 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. QC209 Supplier R&D Systems™ Supplier No. QC209

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with catalog number DTSE100

For use as quantitative controls for the determination of cytokine concentrations in biological fluids. Concentrations have been assigned using R&D Systems™’ Quantikine™ ELISA kits. Controls are prepared in a buffered protein base with preservatives. They contain recombinant human cytokines at low, medium and high concentrations. Controls are supplied lyophilized.

Specifications

Content And Storage Store the unopened product at 2-8°C. Do not use past expiration date.
Gene ID (Entrez) 5054
Product Line R&D Systems
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.