Learn More
Description
Atrial Natriuretic peptide (ANP) is synthesized as 151 aa preproprotein. Proprotein contains cardiodilatin (aa 26 - 92) and atrial natriuretic factor (ANF; aa 124 - 151) which become active after processing by serine protease corin. ANP is primarily synthesized in heart and is secreted in response to atrial distention. ANF circulates in plasma, eliciting natriuretic, diuretic, vasorelaxant and antimitogenic effects directed toward reduction of body fluid and blood pressure regulation. ANF acts through cGMP-coupled NP receptor A. Human ANP and ANF share 84% and 96% aa identity with mouse ANP and ANF, respectively.
- Storage: Store unopened product at 2 to 8°C. Do not use past expiration date.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Type | Control Set for ELISA Kit |
| Gene ID (Entrez) | 4878 (Human); 230899 (Mouse); 24602 (Rat) |
| Intended Use | For quantitative measurement of Atrial Natriuretic peptide (ANP) levels |
| Product Line | R&D Systems |
| Product Name | Immunoassay Control Set 905 |
| Source | Human |
| Target Gene | Atrial Natriuretic peptide |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
