Learn More
Description
- For use as quantitative controls for the determination of cytokine concentrations in biological fluids
- Concentrations have been assigned using R&D Systems’ Quantikine™ kits
- Controls are prepared in diluted serum with preservatives
- They contain recombinant human cytokines at low, medium and high concentrations
- Controls are supplied lyophilized
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Type | Quantikine Immunoassay Control Set 739 Human Myeloperoxidase |
| Gene ID (Entrez) | 4353 (Human); 17523 (Mouse); 303413 (Rat) |
| Intended Use | Control |
| Product Line | R&D Systems |
| Quantity | 1 Each |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
