Learn More
Description
Insulin-like growth factor (IGF)-I (also known as somatomedin C and somatomedin A), IGF-II (multiplication stimulating activity or MSA) belongs to family of insulin-like growth factors that are structurally homologous to proinsulin. Mature IGF-I and IGF-II share approximately 70% sequence identity. Both IGF-I and IGF-II are expressed in many tissues, cell types, may have autocrine, paracrine, endocrine functions. Mature IGF-I and IGF-II are highly conserved between human, bovine, porcine proteins (100% identity), exhibits cross-species activity. The IGFL (insulin-like growth factor-like) family includes four small (∼11 kDa), probably secreted family members in humans, one in mouse. This family shares A and B chain cysteine motifs with IGF superfamily, and has additional cysteine motif within uncleaved region corresponding to C peptide of IGF family.
- Storage: Store unopened product at 2 to 8°C. Do not use past expiration date.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Type | Control Set for ELISA Kit |
| Gene ID (Entrez) | 3481 (Human); 16002 (Mouse) |
| Intended Use | For quantitative measurement of Insulin-like Growth Factor II protein levels |
| Product Line | R&D Systems |
| Product Name | Immunoassay Control Set 930 |
| Source | Human |
| Target Gene | Insulin-like growth factor (IGF)-I |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
