Learn More
Description
Vitamin D binding protein (VitD BP), also known as DBP and Gc-globulin, is 58 kDa glycoprotein that circulates at high concentration in serum and serves as carrier protein for vitamin D. The transport of vitamin D by VitD BP is important for function of wide variety of tissues. VitD BP binds both 25(OH) and hormonally active 1,25(OH)2 forms of vitamin D. VitD BP is primarily expressed in hepatocytes and to lesser extent in kidney. It delivers vitamin D into cells by Megalin-mediated endocytosis. A selectively deglycosylated form of VitD BP known as macrophage activating factor (MAF) blocks angiogenic effects of FGF basic, VEGF, and Angiopoietin 2. VitD BP enhances chemotaxis of monocytes and neutrophils to activated complement component C5a or C5a des Arg (a C-terminally processed form of C5a). The chemotactic cofactor property of VitD BP is eliminated by binding to 1,25(OH)2 vitamin D, but it is not altered by binding to 25(OH) vitamin D or actin.
- Storage: Store unopened product at 2 to 8°C. Do not use past expiration date.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Type | Control Set for ELISA Kit |
| Gene ID (Entrez) | 2638 (Human); 14473 (Mouse); 24384 (Rat) |
| Intended Use | For quantitative measurement of vitamin D binding protein levels |
| Product Line | R&D Systems |
| Product Name | Immunoassay Control Set 925 |
| Source | Human |
| Target Gene | Vit D binding protein |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
