Learn More
Description
IL-17 family is comprised of at least six proinflammatory cytokines that share conserved cysteine-knot structure but diverge at N-terminus. IL-17 family members are glycoproteins secreted as dimers that induce local cytokine production, recruit granulocytes to sites of inflammation. IL-17 is induced by IL-15, IL-23, mainly in activated CD4+ T cells distinct from Th1 or Th2 cells. IL-17F is most homologous to IL-17, but is induced only by IL-23 in activated monocytes. IL-17B binds IL-17B receptor, but not IL-17 receptor; it is most homologous with IL-17D, which is expressed by resting CD4+ T cells, CD19+ B cells. IL-17E is mainly produced by Th2 cells, recruits eosinophils to lung tissue. IL-17C has very restricted expression pattern but has been detected in adult prostate fetal kidney libraries.
- Storage: Store unopened product at 2 to 8°C. Do not use past expiration date.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Type | Control Set of Analytes |
| Gene ID (Entrez) | 3558 (Human); 16183 (Mouse); 116562 (Rat); 396868 (Porcine); 280822 (Bovine); 403989 (Canine); 100034204 (Equine); 751114 (Feline); 100302458 (Rabbit) |
| Product Line | R&D Systems |
| Source | Human |
| Target Gene | IL-2 |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
